Ethyl 5-bromo-2,4-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

50159-500MG 50159-1G 50159-5G 50159-25G
77328-136EA 178.19 USD
77328-136 77346-648 77364-468 77386-746
Ethyl 5-bromo-2,4-dichlorobenzoate ≥97%
Ethyl 5-bromo-2,4-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD24107147
CAS Number: 862243-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR