(6-Bromo-2-chloro-3-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

50155-500MG 50155-1G 50155-5G
77338-652EA 507.79 USD
77338-652 77351-702 77375-966
(6-Bromo-2-chloro-3-fluorophenyl)methanol ≥97%
(6-Bromo-2-chloro-3-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD31637603
CAS Number: 2092620-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR