Ethyl 4-(benzyloxy)-3-fluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

50152-500MG 50152-1G 50152-5G
77337-862EA 449.08 USD
77337-862 77351-698 77375-962
Ethyl 4-(benzyloxy)-3-fluoro-2-methylbenzoate ≥97%
Ethyl 4-(benzyloxy)-3-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₇FO₃
MW: 288.313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403948
CAS Number: 2624417-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR