Methyl 4-(benzyloxy)-3-fluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

50151-500MG 50151-1G 50151-5G
77338-134EA 466.59 USD
77338-134 77351-696 77375-960
Methyl 4-(benzyloxy)-3-fluoro-2-methylbenzoate ≥97%
Methyl 4-(benzyloxy)-3-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₅FO₃
MW: 274.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403947
CAS Number: 2624417-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR