4-(Benzyloxy)-3-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

50150-500MG 50150-1G 50150-5G
77337-006EA 404.79 USD
77337-006 77351-694 77375-958
4-(Benzyloxy)-3-fluoro-2-methylbenzoic acid ≥97%
4-(Benzyloxy)-3-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD32902866
CAS Number: 2586127-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR