4-(Benzyloxy)-3-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

50148-500MG 50148-1G 50148-5G
77335-810EA 366.68 USD
77335-810 77351-690 77375-954
4-(Benzyloxy)-3-fluoro-2-methylbenzaldehyde ≥97%
4-(Benzyloxy)-3-fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD32902865
CAS Number: 2586126-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR