1-(Benzyloxy)-2-fluoro-4-iodo-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

50147-500MG 50147-1G 50147-5G
77336-394EA 387.28 USD
77336-394 77351-688 77375-952
1-(Benzyloxy)-2-fluoro-4-iodo-3-methylbenzene ≥95%
1-(Benzyloxy)-2-fluoro-4-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂FIO
MW: 342.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902864
CAS Number: 2586126-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR