(4-(Benzyloxy)-3-fluoro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

50146-500MG 50146-1G 50146-5G
77328-134EA 318.27 USD
77328-134 77351-686 77375-950
(4-(Benzyloxy)-3-fluoro-2-methylphenyl)(methyl)sulfane ≥97%
(4-(Benzyloxy)-3-fluoro-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₅H₁₅FOS
Storage Temperature: Ambient
MDL Number: MFCD32902863
CAS Number: 2586126-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR