(2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
50143-500MG
50143-1G
50143-5G
50143-25G
77328-130EA
251.32
USD
77328-130
77351-684
77364-464
77386-742
(2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol ≥97%
(2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BF₂O₃ MW: 270.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31631672 CAS Number: 2055257-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...