Methyl 3-fluoro-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

50131-500MG 50131-1G 50131-5G 50131-25G
77328-128EA 162.74 USD
77328-128 77346-080 77364-462 77386-740
Methyl 3-fluoro-5-(trifluoromethyl)benzoate ≥97%
Methyl 3-fluoro-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203707
CAS Number: 773873-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR