(3-Methoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

50115-500MG 50115-1G 50115-5G 50115-25G
77328-126EA 111.24 USD
77328-126 77343-304 77364-460 77375-948
(3-Methoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(3-Methoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₉F₃OS
MW: 222.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489033
CAS Number: 1823046-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR