N,1-Dimethyl-1H-pyrazole-5-carboxamide ≥97%
Supplier: AOBChem USA
|
Warning
|
50099-500MG
50099-1G
50099-5G
50099-25G
77328-124EA
206
USD
77328-124
77351-682
77364-458
77386-738
N,1-Dimethyl-1H-pyrazole-5-carboxamide ≥97%
N,1-Dimethyl-1H-pyrazole-5-carboxamide
Precaución: for Research Use Only
Formula:
C₆H₉N₃O MW: 139.155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08588250 CAS Number: 955554-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...