N-Cyclopropyl-1-methyl-1H-pyrazole-5-carboxamide ≥97%

Supplier: AOBChem USA

Warning

50098-500MG 50098-1G 50098-5G 50098-25G
77328-122EA 212.18 USD
77328-122 77351-680 77364-456 77386-736
N-Cyclopropyl-1-methyl-1H-pyrazole-5-carboxamide ≥97%
N-Cyclopropyl-1-methyl-1H-pyrazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₈H₁₁N₃O
MW: 165.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20359495
CAS Number: 1052622-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR