(1-Methyl-1H-pyrazol-5-yl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

50097-500MG 50097-1G 50097-5G 50097-25G
77328-120EA 122.57 USD
77328-120 77343-994 77364-454 77375-946
(1-Methyl-1H-pyrazol-5-yl)(morpholino)methanone ≥97%
(1-Methyl-1H-pyrazol-5-yl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O₂
MW: 195.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08071967
CAS Number: 728003-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR