Ethyl 2,6-dichloro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

50022-500MG 50022-1G 50022-5G 50022-25G
77328-116EA 138.02 USD
77328-116 77344-652 77364-450 77386-734
Ethyl 2,6-dichloro-3-methylbenzoate ≥97%
Ethyl 2,6-dichloro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
MW: 233.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786811
CAS Number: 1478503-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR