5-Bromo-2-phenoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

50007-500MG 50007-1G 50007-5G 50007-25G
77328-114EA 211.15 USD
77328-114 77351-676 77364-448 77386-732
5-Bromo-2-phenoxybenzonitrile ≥97%
5-Bromo-2-phenoxybenzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₈BrNO
MW: 274.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756668
CAS Number: 337965-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR