4-(3-Bromo-4-methylphenoxy)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

50006-500MG 50006-1G 50006-5G 50006-25G
77328-112EA 120.51 USD
77328-112 77343-732 77364-446 77375-940
4-(3-Bromo-4-methylphenoxy)benzonitrile ≥97%
4-(3-Bromo-4-methylphenoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrNO
MW: 288.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902860
CAS Number: 132529-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR