4-(4-Bromo-3-methylphenoxy)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

50005-500MG 50005-1G 50005-5G 50005-25G
77328-110EA 150.38 USD
77328-110 77345-348 77364-444 77386-730
4-(4-Bromo-3-methylphenoxy)benzonitrile ≥95%
4-(4-Bromo-3-methylphenoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrNO
MW: 288.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25304402
CAS Number: 947162-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR