2-Bromo-1-ethoxy-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

49988-500MG 49988-1G 49988-5G 49988-25G
77328-108EA 222.48 USD
77328-108 77351-674 77364-442 77386-728
2-Bromo-1-ethoxy-3-(trifluoromethyl)benzene ≥97%
2-Bromo-1-ethoxy-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD28738497
CAS Number: 1089192-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR