Ethyl 2-chloro-4-methoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49987-500MG 49987-1G 49987-5G
77339-360EA 582.98 USD
77339-360 77351-672 77375-938
Ethyl 2-chloro-4-methoxy-5-methylbenzoate ≥97%
Ethyl 2-chloro-4-methoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756666
CAS Number: 2383777-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR