(2-Chloro-4-methoxy-5-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

49984-500MG 49984-1G 49984-5G
77339-126EA 553.11 USD
77339-126 77351-668 77375-934
(2-Chloro-4-methoxy-5-methylphenyl)methanol ≥95%
(2-Chloro-4-methoxy-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MW: 186.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22052372
CAS Number: 84657-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR