2-Chloro-4-methoxy-5-methylphenol ≥95%

Supplier: AOBChem USA

Warning

49982-500MG 49982-1G 49982-5G
77340-224EA 843.57 USD
77340-224 77351-664 77375-930
2-Chloro-4-methoxy-5-methylphenol ≥95%
2-Chloro-4-methoxy-5-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756664
CAS Number: 1600530-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR