2'-Fluoro-5'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
49977-500MG
49977-1G
49977-5G
49977-25G
77328-104EA
198.79
USD
77328-104
77351-658
77364-440
77386-726
2'-Fluoro-5'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Fluoro-5'-methoxy-[1,1'-biphenyl]-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₁FO₂ MW: 230.234 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32902859 CAS Number: 1515244-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...