1,3-Dibromo-2-fluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

49968-500MG 49968-1G 49968-5G 49968-25G
77328-100EA 229.69 USD
77328-100 77351-656 77364-436 77386-724
1,3-Dibromo-2-fluoro-5-iodobenzene ≥97%
1,3-Dibromo-2-fluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂FI
Storage Temperature: Ambient
MDL Number: MFCD28784815
CAS Number: 1807033-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR