1,2-Dibromo-4-chloro-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49964-500MG 49964-1G 49964-5G
77328-098EA 348.14 USD
77328-098 77351-654 77375-922
1,2-Dibromo-4-chloro-5-methoxybenzene ≥97%
1,2-Dibromo-4-chloro-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂ClO
MW: 300.375 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31725452
CAS Number: 174913-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR