2-Bromo-5-chloro-4-fluoroiodobenzene ≥95%

Supplier: AOBChem USA

Warning

49960-500MG 49960-1G 49960-5G 49960-25G
77328-096EA 236.9 USD
77328-096 77351-652 77364-434 77386-722
2-Bromo-5-chloro-4-fluoroiodobenzene ≥95%
2-Bromo-5-chloro-4-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
Storage Temperature: Ambient
MDL Number: MFCD18089307
CAS Number: 1263377-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR