7-Chloro-5-iodo-1H-indazole ≥97%
Supplier: AOBChem USA
|
Warning
|
49949-500MG
49949-1G
49949-5G
49949-25G
77328-094EA
199.82
USD
77328-094
77351-650
77364-432
77386-720
7-Chloro-5-iodo-1H-indazole ≥97%
7-Chloro-5-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄ClIN₂ MW: 278.478 g/mol Storage Temperature: Ambient |
CAS Number:
1000343-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...