Isopropyl 5-bromo-2-chloronicotinate ≥95%

Supplier: AOBChem USA

Warning

49947-500MG 49947-1G 49947-5G 49947-25G
77328-092EA 242.05 USD
77328-092 77351-648 77364-430 77386-718
Isopropyl 5-bromo-2-chloronicotinate ≥95%
Isopropyl 5-bromo-2-chloronicotinate

Precaución: for Research Use Only

Formula: C₉H₉BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD18841837
CAS Number: 1340280-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR