Methyl 2,4-dichloro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49940-1G 49940-5G 49940-25G
77341-420EA 85.49 USD
77341-420 77364-428 77375-920
Methyl 2,4-dichloro-3-methylbenzoate ≥97%
Methyl 2,4-dichloro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28786818
CAS Number: 83277-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR