2-Fluoro-3-iodo-6-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

49860-500MG 49860-1G 49860-5G 49860-25G
77328-082EA 165.83 USD
77328-082 77346-164 77364-420 77386-710
2-Fluoro-3-iodo-6-methoxyphenol ≥97%
2-Fluoro-3-iodo-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD32902854
CAS Number: 2385731-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR