Ethyl 2,6-dichloro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

49854-500MG 49854-1G 49854-5G 49854-25G
77328-080EA 225.57 USD
77328-080 77351-638 77364-416 77386-708
Ethyl 2,6-dichloro-3-methoxybenzoate ≥97%
Ethyl 2,6-dichloro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28360054
CAS Number: 90887-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR