3-(Benzyloxy)-2,6-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49849-500MG 49849-1G 49849-5G
77337-880EA 450.11 USD
77337-880 77351-636 77375-914
3-(Benzyloxy)-2,6-dichlorobenzaldehyde ≥97%
3-(Benzyloxy)-2,6-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28360136
CAS Number: 1439373-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR