Thiomorpholino(2,4,6-trichlorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

49840-1G 49840-5G 49840-25G
77341-386EA 83.43 USD
77341-386 77364-414 77375-908
Thiomorpholino(2,4,6-trichlorophenyl)methanone ≥97%
Thiomorpholino(2,4,6-trichlorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₃NOS
Storage Temperature: Ambient
MDL Number: MFCD34756649
CAS Number: 2807455-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR