2,4,6-Trichlorobenzamide ≥95%

Supplier: AOBChem USA

Warning

49834-1G 49834-5G 49834-25G
77341-548EA 92.7 USD
77341-548 77364-408 77375-902
2,4,6-Trichlorobenzamide ≥95%
2,4,6-Trichlorobenzamide

Precaución: for Research Use Only

Formula: C₇H₄Cl₃NO
Storage Temperature: Ambient
MDL Number: MFCD11044015
CAS Number: 23400-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR