Methyl 3-fluoro-2-methoxy-6-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
49828-500MG
49828-1G
49828-5G
77328-076EA
338.87
USD
77328-076
77351-628
77375-898
Methyl 3-fluoro-2-methoxy-6-methylbenzoate ≥97%
Methyl 3-fluoro-2-methoxy-6-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁FO₃ MW: 198.191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34530028 CAS Number: 2112520-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...