Ethyl 3-fluoro-2-methoxy-6-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49827-500MG 49827-1G 49827-5G
77335-400EA 353.29 USD
77335-400 77351-626 77375-896
Ethyl 3-fluoro-2-methoxy-6-methylbenzoate ≥97%
Ethyl 3-fluoro-2-methoxy-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530027
CAS Number: 913723-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR