3-Fluoro-6-methyl-2-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49822-500MG 49822-1G 49822-5G 49822-25G
77328-070EA 278.1 USD
77328-070 77351-622 77364-402 77386-702
3-Fluoro-6-methyl-2-propoxybenzoic acid ≥97%
3-Fluoro-6-methyl-2-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530025
CAS Number: 2734774-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR