4-Chloro-2-fluoro-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

49814-500MG 49814-1G 49814-5G 49814-25G
77328-066EA 280.16 USD
77328-066 77351-618 77364-398 77386-698
4-Chloro-2-fluoro-[1,1'-biphenyl]-3-carboxylic acid ≥97%
4-Chloro-2-fluoro-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD33403944
CAS Number: 2624417-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR