4-Chloro-2-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

49812-500MG 49812-1G 49812-5G
77338-180EA 468.65 USD
77338-180 77351-616 77375-894
4-Chloro-2-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%
4-Chloro-2-fluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈ClFO
MW: 234.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701627
CAS Number: 1214368-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR