4-Chloro-2-fluoro-3-iodo-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

49811-500MG 49811-1G 49811-5G
77335-538EA 357.41 USD
77335-538 77351-614 77375-892
4-Chloro-2-fluoro-3-iodo-1,1'-biphenyl ≥97%
4-Chloro-2-fluoro-3-iodo-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇ClFI
MW: 332.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403943
CAS Number: 2624417-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR