(4-Chloro-2-fluoro-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

49810-500MG 49810-1G 49810-5G
77336-066EA 375.95 USD
77336-066 77351-612 77375-890
(4-Chloro-2-fluoro-[1,1'-biphenyl]-3-yl)(methyl)sulfane ≥95%
(4-Chloro-2-fluoro-[1,1'-biphenyl]-3-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClFS
MW: 252.735 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403942
CAS Number: 2624417-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR