(6-Chloro-2-fluoro-3-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49807-500MG 49807-1G 49807-5G 49807-25G
77328-064EA 120.51 USD
77328-064 77343-728 77364-396 77375-888
(6-Chloro-2-fluoro-3-iodophenyl)(methyl)sulfane ≥97%
(6-Chloro-2-fluoro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅ClFIS
MW: 302.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403941
CAS Number: 2635937-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR