1-Chloro-3-fluoro-4-iodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

49806-500MG 49806-1G 49806-5G 49806-25G
77328-062EA 280.16 USD
77328-062 77351-610 77364-394 77386-696
1-Chloro-3-fluoro-4-iodo-2-methylbenzene ≥97%
1-Chloro-3-fluoro-4-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751676
CAS Number: 1511863-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR