1-(6-Chloro-2-fluoro-3-iodophenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
49805-500MG
49805-1G
49805-5G
49805-25G
77328-060EA
184.37
USD
77328-060
77347-154
77364-392
77386-694
1-(6-Chloro-2-fluoro-3-iodophenyl)ethanone ≥97%
1-(6-Chloro-2-fluoro-3-iodophenyl)ethanone
Precaución: for Research Use Only
Formula:
C₈H₅ClFIO MW: 298.481 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34530023 CAS Number: 2385234-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...