(4-Chloro-2-fluoro-3-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

49802-500MG 49802-1G 49802-5G
77328-058EA 341.96 USD
77328-058 77351-608 77375-886
(4-Chloro-2-fluoro-3-methylphenyl)(methyl)sulfane ≥95%
(4-Chloro-2-fluoro-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752237
CAS Number: 1804878-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR