1-Chloro-3-fluoro-4-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

49796-500MG 49796-1G 49796-5G
77339-850EA 660.23 USD
77339-850 77351-606 77375-882
1-Chloro-3-fluoro-4-methoxy-2-methylbenzene ≥97%
1-Chloro-3-fluoro-4-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MW: 174.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751739
CAS Number: 1807026-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR