(6-Chloro-2-fluoro-3-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

49794-500MG 49794-1G 49794-5G
77339-704EA 615.94 USD
77339-704 77351-602 77375-878
(6-Chloro-2-fluoro-3-methoxyphenyl)(methyl)sulfane ≥95%
(6-Chloro-2-fluoro-3-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34530021
CAS Number: 2707341-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR