(3-Bromo-2-chloro-6-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49766-1G 49766-5G 49766-25G
77340-698EA 60.77 USD
77340-698 77364-388 77375-876
(3-Bromo-2-chloro-6-fluorophenyl)(methyl)sulfane ≥97%
(3-Bromo-2-chloro-6-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrClFS
MW: 255.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403938
CAS Number: 2166631-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR