(3-Fluoro-2,4-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

49764-500MG 49764-1G 49764-5G 49764-25G
77328-056EA 234.84 USD
77328-056 77351-598 77364-386 77386-692
(3-Fluoro-2,4-dimethylphenyl)(methyl)sulfane ≥97%
(3-Fluoro-2,4-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FS
MW: 170.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794630
CAS Number: 1806332-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR