3-Fluoro-2-hydroxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49759-500MG 49759-1G 49759-5G 49759-25G
77328-054EA 95.79 USD
77328-054 77342-552 77364-384 77375-872
3-Fluoro-2-hydroxy-5-methylbenzoic acid ≥97%
3-Fluoro-2-hydroxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD28626273
CAS Number: 1556316-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR